Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-chloro-6-methylpyridine, 98%

Click to view available options
Quantity:
5g
25g
132606-40-7
C6H5BrClN
206.467
MFCD07375117
DTFBHJWQTDQBEM-UHFFFAOYSA-N
5-bromo-2-chloro-6-methylpyridine,3-bromo-6-chloro-2-picoline,3-bromo-6-chloro-2-methyl-pyridine,5-bromo-2-chloro-6-picoline,2-chloro-5-bromo-6-methylpyridine,2-chloro-6-methyl-5-bromopyridine,pyridine, 3-bromo-6-chloro-2-methyl,2-chloro-5-bromo-6-picoline,pyridine, 3-bromo-6-chloro-2-methyl,pubchem6208
14866674
3-bromo-6-chloro-2-methylpyridine
CC1=C(C=CC(=N1)Cl)Br
132606-40-7
C6H5BrClN
206.467
MFCD07375117
DTFBHJWQTDQBEM-UHFFFAOYSA-N
5-bromo-2-chloro-6-methylpyridine,3-bromo-6-chloro-2-picoline,3-bromo-6-chloro-2-methyl-pyridine,5-bromo-2-chloro-6-picoline,2-chloro-5-bromo-6-methylpyridine,2-chloro-6-methyl-5-bromopyridine,pyridine, 3-bromo-6-chloro-2-methyl,2-chloro-5-bromo-6-picoline,pyridine, 3-bromo-6-chloro-2-methyl,pubchem6208
14866674
3-bromo-6-chloro-2-methylpyridine
CC1=C(C=CC(=N1)Cl)Br
Boiling Point 80°C to 84°C (2mmHg)(literature)
Quantity 5g
Melting Point 46°C to 47°C
Beilstein 4243997
Formula Weight 206.47
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-chloro-6-methylpyridine