Alfa Aesar™ 5-Bromo-2-chloroanisole, 97%

Manufacturer: Alfa Aesar™ H5553306

Quantity 5g

 View more versions of this product

Catalog No. AAH5553306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16817-43-9
    Molecular Formula C7H6BrClO
    Formula Weight 221.48
    MDL Number MFCD00078659
    Chemical Name or Material 5-Bromo-2-chloroanisole

    Specifications

    Formula Weight 221.48
    Melting Point 27° to 28°C
    Boiling Point 240° to 245°C
    Quantity 5g
    Flash Point 113°C (235°F)
    Molecular Formula C7H6BrClO
    MDL Number MFCD00078659
    CAS 16817-43-9
    Chemical Name or Material 5-Bromo-2-chloroanisole
    Assay 97%

    Description

    Applications
    5-Bromo-2-chloroanisole is used to produce 4-chloro-3-methoxy-benzonitrile at the temperature of 110°C. It will need solvent dimethylformamide

    Solubility
    Soluble in water.

    Notes
    Keep container tightly closed. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit