Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-chlorobenzoic acid, 98+%

Manufacturer:  Alfa Aesar™ A1221818

Quantity 50g

 View more versions of this product

Catalog No. AAA1221818


  Check Availability
Add to cart

Specifications

Description

Applications
5-Bromo-2-chlorobenzoic acid was used to evaluate the homogeneity of organic powder. It can be used to produce 2-chloro-5-deuteriobenzoic acid.

Solubility
Soluble in water 2.63 g/L @20°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

21739-92-4
235.47
2691432
C7H4BrClO2
MFCD00002415
5-Bromo-2-chlorobenzoic acid

Specifications

21739-92-4
235.47
157° to 162°C
50g
5-Bromo-2-chlorobenzoic acid
C7H4BrClO2
MFCD00002415
2691432
Odorless
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit