Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chlorobenzotrifluoride, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
445-01-2
C7H3BrClF3
259.45
MFCD00000601
XGOCKBMEZPNDPJ-UHFFFAOYSA-N
5-bromo-2-chlorobenzotrifluoride,4-bromo-1-chloro-2-trifluoromethyl benzene,2-chloro-5-bromobenzotrifluoride,5-bromo-2-chloro-benzenetrifluoride,5-bromo-2-chlorotrifluoride,3-bromo-6-chlorobenzotrifluoride,benzene, 4-bromo-1-chloro-2-trifluoromethyl,5-bromo-2-chloro-alpha,alpha,alpha-trifluorotoluene,2-chloro-5-bromo-trifluorotoluene,4-bromo-1-chloro-2-trifluoromethyl-benzene
67959
4-bromo-1-chloro-2-(trifluoromethyl)benzene
C1=CC(=C(C=C1Br)C(F)(F)F)Cl
445-01-2
C7H3BrClF3
259.45
MFCD00000601
XGOCKBMEZPNDPJ-UHFFFAOYSA-N
5-bromo-2-chlorobenzotrifluoride,4-bromo-1-chloro-2-trifluoromethyl benzene,2-chloro-5-bromobenzotrifluoride,5-bromo-2-chloro-benzenetrifluoride,5-bromo-2-chlorotrifluoride,3-bromo-6-chlorobenzotrifluoride,benzene, 4-bromo-1-chloro-2-trifluoromethyl,5-bromo-2-chloro-alpha,alpha,alpha-trifluorotoluene,2-chloro-5-bromo-trifluorotoluene,4-bromo-1-chloro-2-trifluoromethyl-benzene
67959
4-bromo-1-chloro-2-(trifluoromethyl)benzene
C1=CC(=C(C=C1Br)C(F)(F)F)Cl
Packaging Glass Bottle
Density 1.7400g/mL
Color Yellow
Quantity 25g
Flash Point 81°C
Boiling Point 76.0°C to 81.0°C (11.0mmHg)
Refractive Index 1.5060 to 1.5080
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula BrC6H3(Cl)CF3
Formula Weight 259.44
Physical Form Liquid
Percent Purity 99%
Grade Technical
Chemical Name or Material 5-Bromo-2-chlorobenzotrifluoride, tech., 94%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-149-2