Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-chlorophenol, 98+%

Click to view available options
Quantity:
1g
5g
183802-98-4
C6H4BrClO
207.451
MFCD00672940
UEVFFMZHGNYDKM-UHFFFAOYSA-N
2-chloro-5-bromophenol,5-bromo-2-chloro-phenol,phenol, 5-bromo-2-chloro,pubchem1476,acmc-1bxbv,3-bromo-6-chlorophenol,5-bromo-2-chloro phenol,ksc174k5h
820392
5-bromo-2-chlorophenol
C1=CC(=C(C=C1Br)O)Cl
183802-98-4
C6H4BrClO
207.451
MFCD00672940
UEVFFMZHGNYDKM-UHFFFAOYSA-N
2-chloro-5-bromophenol,5-bromo-2-chloro-phenol,phenol, 5-bromo-2-chloro,pubchem1476,acmc-1bxbv,3-bromo-6-chlorophenol,5-bromo-2-chloro phenol,ksc174k5h
820392
5-bromo-2-chlorophenol
C1=CC(=C(C=C1Br)O)Cl
Melting Point 56°C to 58°C
Quantity 1g
Formula Weight 207.46
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-chlorophenol

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning