Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chloropyridine-3-carboxamide, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
75291-85-9
C6H4BrClN2O
235.47
NFUOTGWWIKJNQL-UHFFFAOYSA-N
5-bromo-2-chloronicotinamide,acmc-209oxs,5-bromo-2-chloro-nicotinamide,3-pyridinecarboxamide, 5-bromo-2-chloro
12629794
5-bromo-2-chloropyridine-3-carboxamide
C1=C(C(=NC=C1Br)Cl)C(=O)N
75291-85-9
C6H4BrClN2O
235.47
NFUOTGWWIKJNQL-UHFFFAOYSA-N
5-bromo-2-chloronicotinamide,acmc-209oxs,5-bromo-2-chloro-nicotinamide,3-pyridinecarboxamide, 5-bromo-2-chloro
12629794
5-bromo-2-chloropyridine-3-carboxamide
C1=C(C(=NC=C1Br)Cl)C(=O)N
Quantity 5g
Melting Point 182.0°C to 183.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 235.47
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-chloropyridine-3-carboxamide

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning