Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chlorotoluene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
54932-72-8
C7H6BrCl
205.48
OZFQMHJKAODEON-UHFFFAOYSA-N
5-bromo-2-chlorotoluene,benzene, 4-bromo-1-chloro-2-methyl,2-chloro-5-bromotoluene,pubchem3604,5-bromo-2-chlorotoulene,acmc-209lk8,ksc493o2d,ozfqmhjkaodeon-uhfffaoysa,attercop-chm at115283
609899
4-bromo-1-chloro-2-methylbenzene
CC1=C(C=CC(=C1)Br)Cl
54932-72-8
C7H6BrCl
205.48
OZFQMHJKAODEON-UHFFFAOYSA-N
5-bromo-2-chlorotoluene,benzene, 4-bromo-1-chloro-2-methyl,2-chloro-5-bromotoluene,pubchem3604,5-bromo-2-chlorotoulene,acmc-209lk8,ksc493o2d,ozfqmhjkaodeon-uhfffaoysa,attercop-chm at115283
609899
4-bromo-1-chloro-2-methylbenzene
CC1=C(C=CC(=C1)Br)Cl
Specific Gravity 1.549
Packaging Glass bottle
Boiling Point 90.0°C to 93.0°C (12.0mmHg)
Color Undesignated
Quantity 25g
Infrared Spectrum Authentic
Flash Point >110°C
Refractive Index 1.5735 to 1.5755
Density 1.5490g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 205.48
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-chlorotoluene
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.