Learn More
5-Bromo-2'-deoxyuridine, 99%, Thermo Scientific Chemicals
Description
5-Bromo-2'-deoxyuridine (BrdU) is a thymidine analog used to label DNA. It is incorporated into newly synthesized DNA in place of thymidine during the S phase of the cell cycle. Cells that were actively proliferating can then be detected by denaturing the DNA and allowing specific antibodies to target the BrdU incorporation. Consequently 5-BrdU is used to study cell signaling and other processes that induce cell proliferation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5-Bromo-2′-deoxyuridine (BrdU) is a thymidine analog used to label DNA. It is incorporated into newly synthesized DNA in place of thymidine during the S phase of the cell cycle. Cells that were actively proliferating can then be detected by denaturing the DNA and allowing specific antibodies to target the BrdU incorporation. Consequently 5-BrdU is used to study cell signaling and other processes that induce cell proliferation.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | ∽190°C (decomposition) |
Quantity | 1 g |
Beilstein | 30395 |
Merck Index | 14,1452 |
Solubility Information | Soluble in water. |
Optical Rotation | +31° (c=1 in 0.1N HCl) |
Formula Weight | 307.1 |
Percent Purity | 99% |
Chemical Name or Material | 5-Bromo-2'-deoxyuridine |
Safety and Handling
Hazard Category | H315-H319-H335-H341 |
Hazard Statement | GHS H Statement H341 Suspected of causing genetic defects. |
Precautionary Statement | P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c |
EINECSNumber | 200-415-9 |
RTECSNumber | YU7350000 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.