Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2'-deoxyuridine, 99%

Manufacturer:  Alfa Aesar™ H27260MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27260MD


  Check Availability
Add to cart

Specifications

Description

Applications
5-Bromo-2 ft.-deoxyuridine (BrdU) is a thymidine analog used to label DNA. It is incorporated into newly synthesized DNA in place of thymidine during the S phase of the cell cycle. Cells that were actively proliferating can then be detected by denaturing the DNA and allowing specific antibodies to target the BrdU incorporation. Consequently 5-BrdU is used to study cell signaling and other processes that induce cell proliferation.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

59-14-3
307.1
30395
5-Bromo-1-(2-deoxy-beta-D-ribofuranosyl)uracil; 5-BrdU
C9H11BrN2O5
MFCD00006529
14,1452
5-Bromo-2'-deoxyuridine

Specifications

59-14-3
307.1
≈190°C
14,1452
250mg
5-Bromo-2'-deoxyuridine
C9H11BrN2O5
MFCD00006529
30395
5-Bromo-1-(2-deoxy-beta-D-ribofuranosyl)uracil; 5-BrdU
+31° (c=1 in 0.1N HCl)
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H341
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YU7350000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit