Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-ethoxypyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
55849-30-4
C7H8BrNO
202.05
WQXZKMUZWPUZGL-UHFFFAOYSA-N
2-ethoxy-5-bromopyridine,3-bromo-6-ethoxypyridine,5-bromo-2-ethoxy-pyridine,pyridine, 5-bromo-2-ethoxy,zlchem 405,pubchem6235,5-bromo-ethoxy-pyridine,acmc-1avox,5-bromo-2-ethoxy pyridine,ksc497o6h
817237
5-bromo-2-ethoxypyridine
CCOC1=NC=C(C=C1)Br
55849-30-4
C7H8BrNO
202.05
WQXZKMUZWPUZGL-UHFFFAOYSA-N
2-ethoxy-5-bromopyridine,3-bromo-6-ethoxypyridine,5-bromo-2-ethoxy-pyridine,pyridine, 5-bromo-2-ethoxy,zlchem 405,pubchem6235,5-bromo-ethoxy-pyridine,acmc-1avox,5-bromo-2-ethoxy pyridine,ksc497o6h
817237
5-bromo-2-ethoxypyridine
CCOC1=NC=C(C=C1)Br
Boiling Point 107°C (33.0 Torr)
Flash Point 103°C
Melting Point 34°C to 37°C
Infrared Spectrum Authentic
Color White to Yellow
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 202.05
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-ethoxypyridine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning