Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-fluorobenzylamine hydrochloride, 97%

Click to view available options
Quantity:
1g
5g
25g
202865-69-8
C7H8BrClFN
240.5
MFCD00143426
KPOKOEWRTGFXDF-UHFFFAOYSA-N
5-bromo-2-fluorobenzylamine hydrochloride,5-bromo-2-fluorophenyl methanamine hydrochloride,5-bromo-2-fluorobenzylamine hcl,5-bromo-2-fluorophenyl methanamine hcl,1-5-bromo-2-fluorophenyl methanamine hydrochloride,pubchem3227,acmc-1cpb5,rarechem al bd 0325,akos bbv-005136,5-bromo-2-fluorobenzyl amine hcl
2724903
(5-bromo-2-fluorophenyl)methanamine;hydrochloride
C1=CC(=C(C=C1Br)CN)F.Cl
202865-69-8
C7H8BrClFN
240.5
MFCD00143426
KPOKOEWRTGFXDF-UHFFFAOYSA-N
5-bromo-2-fluorobenzylamine hydrochloride,5-bromo-2-fluorophenyl methanamine hydrochloride,5-bromo-2-fluorobenzylamine hcl,5-bromo-2-fluorophenyl methanamine hcl,1-5-bromo-2-fluorophenyl methanamine hydrochloride,pubchem3227,acmc-1cpb5,rarechem al bd 0325,akos bbv-005136,5-bromo-2-fluorobenzyl amine hcl
2724903
(5-bromo-2-fluorophenyl)methanamine;hydrochloride
C1=CC(=C(C=C1Br)CN)F.Cl
Melting Point 221°C to 226°C
Quantity 25g
Formula Weight 240.5
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-fluorobenzylamine hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning