Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-fluoropyridine, 99%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
766-11-0
C5H3BrFN
175.99
MFCD01863742
MYUQKYGWKHTRPG-UHFFFAOYSA-N
2-fluoro-5-bromopyridine,3-bromo-6-fluoropyridine,5-bromo-2-fluoro pyridine,5-bromo-2-fluoro-pyridine,pyridine, 5-bromo-2-fluoro,2-fluoro-5-bromo pyridine,pubchem3550,2-fluro-5-bromo-pyridine,acmc-209p4s,bromo-2-fluoropyridine, 5
2783168
5-bromo-2-fluoropyridine
C1=CC(=NC=C1Br)F
766-11-0
C5H3BrFN
175.99
MFCD01863742
MYUQKYGWKHTRPG-UHFFFAOYSA-N
2-fluoro-5-bromopyridine,3-bromo-6-fluoropyridine,5-bromo-2-fluoro pyridine,5-bromo-2-fluoro-pyridine,pyridine, 5-bromo-2-fluoro,2-fluoro-5-bromo pyridine,pubchem3550,2-fluro-5-bromo-pyridine,acmc-209p4s,bromo-2-fluoropyridine, 5
2783168
5-bromo-2-fluoropyridine
C1=CC(=NC=C1Br)F
Specific Gravity 1.72
Flash Point 73°C
Quantity 5g
Boiling Point 170.0°C to 173.0°C
Refractive Index 1.5310 to 1.5330
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Density 1.7200g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 175.99
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 5-Bromo-2-fluoropyridine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physicia

GHS Signal Word: Danger