Alfa Aesar™ 5-Bromo-2-fluorotoluene, 98+%

Manufacturer: Alfa Aesar™ A1072106

Quantity 5g

 View more versions of this product

Catalog No. AAA1072106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51437-00-4
    Molecular Formula C7H6BrF
    Formula Weight 189.03
    MDL Number MFCD00000343
    Beilstein 2242693
    Chemical Name or Material 5-Bromo-2-fluorotoluene

    Specifications

    Formula Weight 189.03
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    Boiling Point 94° to 95°C (50mmHg)
    Quantity 5g
    Flash Point 73°C (163°F)
    MDL Number MFCD00000343
    CAS 51437-00-4
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Molecular Formula C7H6BrF
    Beilstein 2242693
    Chemical Name or Material 5-Bromo-2-fluorotoluene
    Assay ≥98%

    Description

    Applications
    5-Bromo-2-fluorotoluene is used in the preparation of 5-bromo-2-fluorobenzyl bromide via N-bromosuccinimide (NBS) bromination, required for the synthesis of crown bromofluorobenzene and 3-(4-fuoro-3-methylphenyl)acrylamide. It is also used as an intermediates of medicine.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit