Alfa Aesar™ 5-Bromo-2-hydroxy-3-methoxybenzyl alcohol

Manufacturer: Alfa Aesar™ H5056903

Quantity 1g

 View more versions of this product

Catalog No. AAH5056903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 35090-64-3
    Molecular Formula C8H9BrO3
    Formula Weight 233.06
    MDL Number MFCD02081986
    Synonym 4-Bromo-2-hydroxymethyl-6-methoxyphenol
    Chemical Name or Material 5-Bromo-2-hydroxy-3-methoxybenzyl alcohol

    Specifications

    Formula Weight 233.06
    Molecular Formula C8H9BrO3
    MDL Number MFCD02081986
    CAS 35090-64-3
    Synonym 4-Bromo-2-hydroxymethyl-6-methoxyphenol
    Quantity 1g
    Chemical Name or Material 5-Bromo-2-hydroxy-3-methoxybenzyl alcohol

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit