Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-hydroxy-4-methylpyridine, 97%

Click to view available options
Quantity:
5g
25g
164513-38-6
C6H6BrNO
188.024
MFCD03427664
BYGNXKDNIFHZHQ-UHFFFAOYSA-N
5-bromo-2-hydroxy-4-methylpyridine,5-bromo-4-methylpyridin-2-ol,5-bromo-2-hydroxy-4-picoline,5-bromo-4-methyl-2 1h-pyridinone,5-bromo-4-methyl-2-pyridinol,2-hydroxy-4-methyl-5-bromopyridine,5-bromo-4-methyl-pyridin-2-ol,pubchem6564,acmc-209dre,ksc496a8d
819937
5-bromo-4-methyl-1H-pyridin-2-one
CC1=CC(=O)NC=C1Br
164513-38-6
C6H6BrNO
188.024
MFCD03427664
BYGNXKDNIFHZHQ-UHFFFAOYSA-N
5-bromo-2-hydroxy-4-methylpyridine,5-bromo-4-methylpyridin-2-ol,5-bromo-2-hydroxy-4-picoline,5-bromo-4-methyl-2 1h-pyridinone,5-bromo-4-methyl-2-pyridinol,2-hydroxy-4-methyl-5-bromopyridine,5-bromo-4-methyl-pyridin-2-ol,pubchem6564,acmc-209dre,ksc496a8d
819937
5-bromo-4-methyl-1H-pyridin-2-one
CC1=CC(=O)NC=C1Br
Applications
It is used for medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Quantity 25g
Melting Point 198°C to 202°C
Color White
Beilstein 6798531
Formula Weight 188.02
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxy-4-methylpyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning