Alfa Aesar™ 5-Bromo-2-hydroxybenzyl alcohol, 98+%

Manufacturer: Alfa Aesar™ A1234414

Quantity 25g

 View more versions of this product

Catalog No. AAA1234414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2316-64-5
    Molecular Formula C7H7BrO2
    Formula Weight 203.04
    MDL Number MFCD00004618
    Beilstein 509726
    Synonym 5-Bromosalicyl alcohol; Bromosaligenin
    Chemical Name or Material 5-Bromo-2-hydroxybenzyl alcohol

    Specifications

    Formula Weight 203.04
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    CAS 2316-64-5
    Beilstein 509726
    Quantity 25g
    Health Hazard 1 Warning
    Molecular Formula C7H7BrO2
    MDL Number MFCD00004618
    Synonym 5-Bromosalicyl alcohol; Bromosaligenin
    Melting Point 108° to 110°C
    Chemical Name or Material 5-Bromo-2-hydroxybenzyl alcohol
    Assay ≥98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit