Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-hydroxypyridine, 97%

Click to view available options
Quantity:
5g
25g
100g
13466-38-1
C5H4BrNO
173.997
MFCD00234042
NDMZZQRNZFWMEZ-UHFFFAOYSA-N
5-bromo-2-hydroxypyridine,5-bromopyridin-2-ol,2-hydroxy-5-bromopyridine,5-bromopyridin-2 1h-one,5-bromo-2-pyridinol,5-bromo-2 1h-pyridinone,5-bromo-2 1h-pyridone,2 1h-pyridinone, 5-bromo,3-bromo-6-hydroxypyridine,5-bromo-1,2-dihydropyridin-2-one
599528
5-bromo-1H-pyridin-2-one
C1=CC(=O)NC=C1Br
13466-38-1
C5H4BrNO
173.997
MFCD00234042
NDMZZQRNZFWMEZ-UHFFFAOYSA-N
5-bromo-2-hydroxypyridine,5-bromopyridin-2-ol,2-hydroxy-5-bromopyridine,5-bromopyridin-2 1h-one,5-bromo-2-pyridinol,5-bromo-2 1h-pyridinone,5-bromo-2 1h-pyridone,2 1h-pyridinone, 5-bromo,3-bromo-6-hydroxypyridine,5-bromo-1,2-dihydropyridin-2-one
599528
5-bromo-1H-pyridin-2-one
C1=CC(=O)NC=C1Br
Melting Point 178°C to 182°C
Quantity 100g
Beilstein 108751
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxypyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning