Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-iodobenzonitrile, 98+%

Click to view available options
Quantity:
1g
5g
25g
121554-10-7
C7H3BrIN
307.916
MFCD00672950
JIGKPMMZNMQXDL-UHFFFAOYSA-N
benzonitrile, 5-bromo-2-iodo,5-bromo-2-iodobenzenecarbonitrile,2-iodo-5-bromobenzonitrile,2-cyano-5-bromo-1-iodobenzene,5-bromo-2-iodobenzonitrile, 98+%,pubchem3790,acmc-1cir6,5-bromo-2-iodo-benzonitrile,ksc493s3b,benzonitrile,5-bromo-2-iodo
7020874
5-bromo-2-iodobenzonitrile
C1=CC(=C(C=C1Br)C#N)I
121554-10-7
C7H3BrIN
307.916
MFCD00672950
JIGKPMMZNMQXDL-UHFFFAOYSA-N
benzonitrile, 5-bromo-2-iodo,5-bromo-2-iodobenzenecarbonitrile,2-iodo-5-bromobenzonitrile,2-cyano-5-bromo-1-iodobenzene,5-bromo-2-iodobenzonitrile, 98+%,pubchem3790,acmc-1cir6,5-bromo-2-iodo-benzonitrile,ksc493s3b,benzonitrile,5-bromo-2-iodo
7020874
5-bromo-2-iodobenzonitrile
C1=CC(=C(C=C1Br)C#N)I
Quantity 1g
Melting Point 120°C to 122°C
UN Number UN3276
Beilstein 4860282
Formula Weight 307.92
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-iodobenzonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.