Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-iodobenzonitrile, 98+%

Click to view available options
Quantity:
1g
5g
25g
121554-10-7
C7H3BrIN
307.916
MFCD00672950
JIGKPMMZNMQXDL-UHFFFAOYSA-N
benzonitrile, 5-bromo-2-iodo,5-bromo-2-iodobenzenecarbonitrile,2-iodo-5-bromobenzonitrile,2-cyano-5-bromo-1-iodobenzene,5-bromo-2-iodobenzonitrile, 98+%,pubchem3790,acmc-1cir6,5-bromo-2-iodo-benzonitrile,ksc493s3b,benzonitrile,5-bromo-2-iodo
7020874
5-bromo-2-iodobenzonitrile
C1=CC(=C(C=C1Br)C#N)I
121554-10-7
C7H3BrIN
307.916
MFCD00672950
JIGKPMMZNMQXDL-UHFFFAOYSA-N
benzonitrile, 5-bromo-2-iodo,5-bromo-2-iodobenzenecarbonitrile,2-iodo-5-bromobenzonitrile,2-cyano-5-bromo-1-iodobenzene,5-bromo-2-iodobenzonitrile, 98+%,pubchem3790,acmc-1cir6,5-bromo-2-iodo-benzonitrile,ksc493s3b,benzonitrile,5-bromo-2-iodo
7020874
5-bromo-2-iodobenzonitrile
C1=CC(=C(C=C1Br)C#N)I
Melting Point 120°C to 122°C
Sensitivity Light sensitive
Quantity 5g
UN Number UN3276
Beilstein 4860282
Formula Weight 307.92
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-iodobenzonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 601-788-4

TSCA : No

Recommended Storage : Ambient temperatures