Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-iodotoluene, 98+%

Click to view available options
Quantity:
5g
25g
100g
116632-39-4
C7H6BrI
296.933
MFCD00060664
GHTUADBHTFHMNI-UHFFFAOYSA-N
5-bromo-2-iodotoluene,1-bromo-4-iodo-5-methylbenzene,benzene, 4-bromo-1-iodo-2-methyl,bromoiodotoluene,pubchem3793,2-iodo-5-bromo-toluene,5-bromo-2-iodo toluene,5-bromo-2-iodotoluene,,4-bromo-2-methyliodobenzene,4-bromo-2-methyl-iodobenzene
2724601
4-bromo-1-iodo-2-methylbenzene
CC1=C(C=CC(=C1)Br)I
116632-39-4
C7H6BrI
296.933
MFCD00060664
GHTUADBHTFHMNI-UHFFFAOYSA-N
5-bromo-2-iodotoluene,1-bromo-4-iodo-5-methylbenzene,benzene, 4-bromo-1-iodo-2-methyl,bromoiodotoluene,pubchem3793,2-iodo-5-bromo-toluene,5-bromo-2-iodo toluene,5-bromo-2-iodotoluene,,4-bromo-2-methyliodobenzene,4-bromo-2-methyl-iodobenzene
2724601
4-bromo-1-iodo-2-methylbenzene
CC1=C(C=CC(=C1)Br)I
Applications
5-Bromo-2-iodotoluene is used as a medical intermediate and in organic synthesis.

Notes
Light sensitive. Incompatible with Strong oxidizing agents, light.
Flash Point >110°C (230°F)
Boiling Point 262°C to 265°C
Quantity 100g
Beilstein 2325157
Formula Weight 296.93
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-iodotoluene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning