Alfa Aesar™ 5-Bromo-2-methoxy-beta-nitrostyrene, 97%

Manufacturer: Alfa Aesar™ B2482406

Quantity 5g

 View more versions of this product

Catalog No. AAB2482406

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 175205-14-8
    Molecular Formula C9H8BrNO3
    Formula Weight 258.07
    MDL Number MFCD00068087
    Chemical Name or Material 5-Bromo-2-methoxy-beta-nitrostyrene

    Specifications

    Formula Weight 258.07
    Molecular Formula C9H8BrNO3
    MDL Number MFCD00068087
    CAS 175205-14-8
    Melting Point 120° to 122°C
    Quantity 5g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 5-Bromo-2-methoxy-beta-nitrostyrene
    Assay 97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit