Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-methoxy-beta-nitrostyrene, 97%

Click to view available options
Quantity:
1g
5g
175205-14-8
C9H8BrNO3
258.071
MFCD00068087
IAHCHTBHTXVKTC-SNAWJCMRSA-N
4-bromo-1-methoxy-2-2-nitrovinyl benzene,4-bromo-1-methoxy-2-e-2-nitroethenyl benzene,5-bromo-2-methoxy-omega-nitrostyrene,2-1e-2-nitrovinyl-4-bromo-1-methoxybenzene,benzene,4-bromo-1-methoxy-2-2-nitroethenyl,beta-nitro-2-methoxy-5-bromostyrene
5358770
4-bromo-1-methoxy-2-[(E)-2-nitroethenyl]benzene
COC1=C(C=C(C=C1)Br)C=C[N+](=O)[O-]
175205-14-8
C9H8BrNO3
258.071
MFCD00068087
IAHCHTBHTXVKTC-SNAWJCMRSA-N
4-bromo-1-methoxy-2-2-nitrovinyl benzene,4-bromo-1-methoxy-2-e-2-nitroethenyl benzene,5-bromo-2-methoxy-omega-nitrostyrene,2-1e-2-nitrovinyl-4-bromo-1-methoxybenzene,benzene,4-bromo-1-methoxy-2-2-nitroethenyl,beta-nitro-2-methoxy-5-bromostyrene
5358770
4-bromo-1-methoxy-2-[(E)-2-nitroethenyl]benzene
COC1=C(C=C(C=C1)Br)C=C[N+](=O)[O-]
Quantity 1g
Melting Point 120°C to 122°C
Formula Weight 258.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methoxy-beta-nitrostyrene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning