Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methoxybenzyl alcohol, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
80866-82-6
C8H9BrO2
217.062
MFCD00004630
HAOOGRLZVQWDBY-UHFFFAOYSA-N
5-bromo-2-methoxybenzyl alcohol,5-bromo-2-methoxyphenyl methanol,5-bromo-2-methoxybenzylalcohol,benzenemethanol, 5-bromo-2-methoxy,4-bromo-2-hydroxymethyl anisole,5-bromo-2-methoxy-phenyl-methanol,2-methoxy-5-bromobenzyl alcohol,5-bromo-o-anisyl alcohol,4-bromo-hydroxymethyl anisole,benzenemethanol,5-bromo-2-methoxy
612298
(5-bromo-2-methoxyphenyl)methanol
COC1=C(C=C(C=C1)Br)CO
80866-82-6
C8H9BrO2
217.062
MFCD00004630
HAOOGRLZVQWDBY-UHFFFAOYSA-N
5-bromo-2-methoxybenzyl alcohol,5-bromo-2-methoxyphenyl methanol,5-bromo-2-methoxybenzylalcohol,benzenemethanol, 5-bromo-2-methoxy,4-bromo-2-hydroxymethyl anisole,5-bromo-2-methoxy-phenyl-methanol,2-methoxy-5-bromobenzyl alcohol,5-bromo-o-anisyl alcohol,4-bromo-hydroxymethyl anisole,benzenemethanol,5-bromo-2-methoxy
612298
(5-bromo-2-methoxyphenyl)methanol
COC1=C(C=C(C=C1)Br)CO
Melting Point 68°C to 73°C
Infrared Spectrum Authentic
Color White
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula BrC6H3(OCH3)CH2OH
Formula Weight 217.06
Physical Form Needles
Percent Purity ≥96.0%
Chemical Name or Material 5-Bromo-2-methoxybenzyl alcohol, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 279-586-7