Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methoxypyridine-3-carboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
103058-87-3
C7H6BrNO2
216.04
VRNOWEKCASDTFG-UHFFFAOYSA-N
5-bromo-2-methoxy-pyridine-3-carbaldehyde,5-bromo-2-methoxynicotinaldehyde,5-bromo-2-methoxypyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 5-bromo-2-methoxy,5-bromo-2-methoxypyridin-3-carboxaldehyde,5-bromo-2-methoxy-3-pyridinecarboxaldehyde,pubchem9207,abbypharma ap-10-5904,3-pyridinecarboxaldehyde,5-bromo-2-methoxy,5-bromo-2-methoxypyridine-3-carboxaldehyde 1g
1516403
5-bromo-2-methoxypyridine-3-carbaldehyde
COC1=NC=C(C=C1C=O)Br
103058-87-3
C7H6BrNO2
216.04
VRNOWEKCASDTFG-UHFFFAOYSA-N
5-bromo-2-methoxy-pyridine-3-carbaldehyde,5-bromo-2-methoxynicotinaldehyde,5-bromo-2-methoxypyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 5-bromo-2-methoxy,5-bromo-2-methoxypyridin-3-carboxaldehyde,5-bromo-2-methoxy-3-pyridinecarboxaldehyde,pubchem9207,abbypharma ap-10-5904,3-pyridinecarboxaldehyde,5-bromo-2-methoxy,5-bromo-2-methoxypyridine-3-carboxaldehyde 1g
1516403
5-bromo-2-methoxypyridine-3-carbaldehyde
COC1=NC=C(C=C1C=O)Br
Melting Point 102.0°C to 104.0°C
Infrared Spectrum Authentic
Quantity 5g
Color White to Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 216.04
Physical Form Powder or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methoxypyridine-3-carboxaldehyde
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Warning