Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methoxypyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
13472-85-0
C6H6BrNO
188.024
MFCD01318952
XADICJHFELMBGX-UHFFFAOYSA-N
2-methoxy-5-bromopyridine,pyridine, 5-bromo-2-methoxy,3-bromo-6-methoxypyridine,5-bromo-2-methoxy-pyridine,2-methyloxy-5-bromopyridine,5-bromo-2-methoxy pyridine,2-methoxy-5-bromo-pyridine,2-methyloxy-5-bromo pyridine,zlchem 355,pubchem2408
2734895
5-bromo-2-methoxypyridine
COC1=NC=C(C=C1)Br
13472-85-0
C6H6BrNO
188.024
MFCD01318952
XADICJHFELMBGX-UHFFFAOYSA-N
2-methoxy-5-bromopyridine,pyridine, 5-bromo-2-methoxy,3-bromo-6-methoxypyridine,5-bromo-2-methoxy-pyridine,2-methyloxy-5-bromopyridine,5-bromo-2-methoxy pyridine,2-methoxy-5-bromo-pyridine,2-methyloxy-5-bromo pyridine,zlchem 355,pubchem2408
2734895
5-bromo-2-methoxypyridine
COC1=NC=C(C=C1)Br
Quantity 5mL
Density 1.4530g/mL
Flash Point 96°C
Infrared Spectrum Authentic
Boiling Point 80.0°C (12.0 mmHg)
Packaging Glass bottle
Specific Gravity 1.453
Refractive Index 1.5540 to 1.5560
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 188.02
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methoxypyridine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning