Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methylaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
39478-78-9
C7H8BrN
186.05
MFCD00800678
RXQNKKRGJJRMKD-UHFFFAOYSA-N
2-amino-4-bromotoluene,5-bromo-o-toluidine,3-bromo-6-methylaniline,5-bromo-2-methyl-aniline,benzenamine, 5-bromo-2-methyl,5-bromo-2-methyl aniline,5-bromo-2-methyl-phenylamine,zlchem 583,pubchem3801,acmc-1ajfs
2734805
5-bromo-2-methylaniline
CC1=C(C=C(C=C1)Br)N
39478-78-9
C7H8BrN
186.05
MFCD00800678
RXQNKKRGJJRMKD-UHFFFAOYSA-N
2-amino-4-bromotoluene,5-bromo-o-toluidine,3-bromo-6-methylaniline,5-bromo-2-methyl-aniline,benzenamine, 5-bromo-2-methyl,5-bromo-2-methyl aniline,5-bromo-2-methyl-phenylamine,zlchem 583,pubchem3801,acmc-1ajfs
2734805
5-bromo-2-methylaniline
CC1=C(C=C(C=C1)Br)N
Boiling Point 139°C
Color Yellow to Brown
Flash Point 110°C
Specific Gravity 1.49
Refractive Index 1.6120 to 1.614
Density 1.4900g/mL
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H3(CH3)NH2
Formula Weight 186.05
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-methylaniline
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 254-467-2