Alfa Aesar™ 5-Bromo-2-methylbenzotrifluoride, 97+%

Manufacturer: Alfa Aesar™ B2343106

Quantity 5g

 View more versions of this product

Catalog No. AAB2343106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 86845-27-4
    Molecular Formula C8H6BrF3
    Formula Weight 239.04
    MDL Number MFCD01631587
    Synonym 4-Bromo-2-(trifluoromethyl)toluene; 4-Methyl-3-(trifluoromethyl)bromobenzene
    Chemical Name or Material 5-Bromo-2-methylbenzotrifluoride

    Specifications

    Formula Weight 239.04
    Molecular Formula C8H6BrF3
    MDL Number MFCD01631587
    CAS 86845-27-4
    Synonym 4-Bromo-2-(trifluoromethyl)toluene; 4-Methyl-3-(trifluoromethyl)bromobenzene
    Quantity 5g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 5-Bromo-2-methylbenzotrifluoride
    Assay ≥97%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit