Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-methylbenzotrifluoride, 97+%

Click to view available options
Quantity:
1g
5g
86845-27-4
C8H6BrF3
239.035
MFCD01631587
IZFVIEYHUOUPPH-UHFFFAOYSA-N
5-bromo-2-methylbenzotrifluoride,4-methyl-3-trifluoromethyl bromobenzene,4-bromo-1-methyl-2-trifluoromethyl benzene,3-bromo-6-methylbenzotrifluoride,4-bromo-2-trifluoromethyl toluene,4-bromo-1-methyl-2-trifluoromethyl-benzene,benzene, 4-bromo-1-methyl-2-trifluoromethyl,benzene,4-bromo-1-methyl-2-trifluoromethyl,ksc657e6d
2775635
4-bromo-1-methyl-2-(trifluoromethyl)benzene
CC1=C(C=C(C=C1)Br)C(F)(F)F
86845-27-4
C8H6BrF3
239.035
MFCD01631587
IZFVIEYHUOUPPH-UHFFFAOYSA-N
5-bromo-2-methylbenzotrifluoride,4-methyl-3-trifluoromethyl bromobenzene,4-bromo-1-methyl-2-trifluoromethyl benzene,3-bromo-6-methylbenzotrifluoride,4-bromo-2-trifluoromethyl toluene,4-bromo-1-methyl-2-trifluoromethyl-benzene,benzene, 4-bromo-1-methyl-2-trifluoromethyl,benzene,4-bromo-1-methyl-2-trifluoromethyl,ksc657e6d
2775635
4-bromo-1-methyl-2-(trifluoromethyl)benzene
CC1=C(C=C(C=C1)Br)C(F)(F)F
Quantity 1g
Formula Weight 239.04
Percent Purity ≥97%
Chemical Name or Material 5-Bromo-2-methylbenzotrifluoride

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning