Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-methylindole, 96%

Click to view available options
Quantity:
1g
5g
1075-34-9
C9H8BrN
210.074
MFCD01863677
BJUZAZKEDCDGRW-UHFFFAOYSA-N
5-bromo-2-methylindole,2-methyl-5-bromoindole,1h-indole, 5-bromo-2-methyl,5-bromo-2-methylindol,pubchem9382,acmc-209xly,5-bromo-2-methyl-indole,2-methyl-5-bromo-1h-indole,1h-indole,5-bromo-2-methyl,5-bromo-2-methylindole
5003968
5-bromo-2-methyl-1H-indole
CC1=CC2=C(N1)C=CC(=C2)Br
1075-34-9
C9H8BrN
210.074
MFCD01863677
BJUZAZKEDCDGRW-UHFFFAOYSA-N
5-bromo-2-methylindole,2-methyl-5-bromoindole,1h-indole, 5-bromo-2-methyl,5-bromo-2-methylindol,pubchem9382,acmc-209xly,5-bromo-2-methyl-indole,2-methyl-5-bromo-1h-indole,1h-indole,5-bromo-2-methyl,5-bromo-2-methylindole
5003968
5-bromo-2-methyl-1H-indole
CC1=CC2=C(N1)C=CC(=C2)Br
Quantity 1g
Melting Point 104°C to 107°C
Color Brown to White
Beilstein 116647
Formula Weight 210.07
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 5-Bromo-2-methylindole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning