Alfa Aesar™ 5-Bromo-2-methylphenol, 95%

Manufacturer: Alfa Aesar™ H2695609

Quantity 10g

 View more versions of this product

Catalog No. AAH2695609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 36138-76-8
    Molecular Formula C7H7BrO
    Formula Weight 187.04
    MDL Number MFCD08061906
    Synonym 5-Bromo-o-cresol
    Chemical Name or Material 5-Bromo-2-methylphenol

    Specifications

    Formula Weight 187.04
    Melting Point 77° to 80°C
    Quantity 10g
    Molecular Formula C7H7BrO
    MDL Number MFCD08061906
    CAS 36138-76-8
    Synonym 5-Bromo-o-cresol
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P337+P313
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
    Chemical Name or Material 5-Bromo-2-methylphenol
    Assay 95%

    GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P337+P313
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit