Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-methylpyrimidine, 98%

Click to view available options
Quantity:
250mg
1g
7752-78-5
C5H5BrN2
173.013
MFCD07375143
NEDJTEXNSTUKHW-UHFFFAOYSA-N
5-bromo-2-methyl-pyrimidine,2-methyl-5-bromopyrimidine,pyrimidine, 5-bromo-2-methyl,5-bromo-2-methyl-1,3-diazine,pubchem15231,acmc-1bm3h,5-bromo-2-methyl pyrimidine
14387744
5-bromo-2-methylpyrimidine
CC1=NC=C(C=N1)Br
7752-78-5
C5H5BrN2
173.013
MFCD07375143
NEDJTEXNSTUKHW-UHFFFAOYSA-N
5-bromo-2-methyl-pyrimidine,2-methyl-5-bromopyrimidine,pyrimidine, 5-bromo-2-methyl,5-bromo-2-methyl-1,3-diazine,pubchem15231,acmc-1bm3h,5-bromo-2-methyl pyrimidine
14387744
5-bromo-2-methylpyrimidine
CC1=NC=C(C=N1)Br
Quantity 1g
Beilstein 108736á
Formula Weight 173.01
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-methylpyrimidine

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : 2° to 8°C