Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-bromo-2-picoline 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
3430-13-5
C6H6BrN
172.025
MFCD00661170
OFKWIQJLYCKDNY-UHFFFAOYSA-N
5-bromo-2-picoline,2-methyl-5-bromopyridine,5-bromo-2-methyl-pyridine,3-bromo-6-methylpyridine,pyridine, 5-bromo-2-methyl,5-bromopicoline,5-bromo-2-methyl pyridine,3-bromo-6-picoline,pubchem4058
1201020
5-bromo-2-methylpyridine
CC1=NC=C(C=C1)Br
3430-13-5
C6H6BrN
172.025
MFCD00661170
OFKWIQJLYCKDNY-UHFFFAOYSA-N
5-bromo-2-picoline,2-methyl-5-bromopyridine,5-bromo-2-methyl-pyridine,3-bromo-6-methylpyridine,pyridine, 5-bromo-2-methyl,5-bromopicoline,5-bromo-2-methyl pyridine,3-bromo-6-picoline,pubchem4058
1201020
5-bromo-2-methylpyridine
CC1=NC=C(C=C1)Br
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 32°C to 36°C
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 172.02
Physical Form Crystals and Chunks
Percent Purity ≥98.5%
Chemical Name or Material 5-Bromo-2-picoline
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.