Alfa Aesar™ 5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, 95%

Manufacturer: Alfa Aesar™ H32316MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH32316MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 749875-16-9
    Molecular Formula C7H3BrF3NO2
    Formula Weight 270
    MDL Number MFCD10687236
    Synonym 5-Bromo-2-(trifluoromethyl)isonicotinic acid
    Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid

    Specifications

    Formula Weight 270
    Molecular Formula C7H3BrF3NO2
    MDL Number MFCD10687236
    CAS 749875-16-9
    Melting Point 235° to 238°C
    Synonym 5-Bromo-2-(trifluoromethyl)isonicotinic acid
    Quantity 250mg
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
    Assay 95%

    Description

    Applications
    5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

    Solubility
    Slightly soluble in water, soluble in organic solvents.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit