Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-2-(trifluoromethyl)pyrimidine, 95%

Click to view available options
Quantity:
250mg
1g
799557-86-1
C5H2BrF3N2
226.984
MFCD09754046
GIFDWXWNFKZVEI-UHFFFAOYSA-N
5-bromo-2-trifluoromethyl pyrimidine,5-bromo-2-trifluoromethyl-pyrimidine,pyrimidine, 5-bromo-2-trifluoromethyl,bromotrifluoromethylpyrimidine,5-bromo-2-trifluoromethyl-pyrimidine,2-trifluoromethyl-5-bromopyrimidine,5-bromo-2-trifluoromethyl-1,3-diazine,2-acetamido-?-methyl-?-thiazolesulfonyl chloride
33697963
5-bromo-2-(trifluoromethyl)pyrimidine
C1=C(C=NC(=N1)C(F)(F)F)Br
799557-86-1
C5H2BrF3N2
226.984
MFCD09754046
GIFDWXWNFKZVEI-UHFFFAOYSA-N
5-bromo-2-trifluoromethyl pyrimidine,5-bromo-2-trifluoromethyl-pyrimidine,pyrimidine, 5-bromo-2-trifluoromethyl,bromotrifluoromethylpyrimidine,5-bromo-2-trifluoromethyl-pyrimidine,2-trifluoromethyl-5-bromopyrimidine,5-bromo-2-trifluoromethyl-1,3-diazine,2-acetamido-?-methyl-?-thiazolesulfonyl chloride
33697963
5-bromo-2-(trifluoromethyl)pyrimidine
C1=C(C=NC(=N1)C(F)(F)F)Br
Color White
Quantity 250mg
Formula Weight 226.98
Physical Form Crystalline
Percent Purity 95%
Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyrimidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning