Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-3-chloro-2-fluoropyridine, 96%

Click to view available options
Quantity:
250mg
1g
5g
38185-56-7
C5H2BrClFN
210.43
MFCD09834365
WXKLHBHJIBAXQH-UHFFFAOYSA-N
2-fluoro-3-chloro-5-bromopyridine,3-bromo-5-chloro-6-fluoropyridine,5-bromo-3-chloro-2-fluoro-pyridine,acmc-1ct80,5-bromo-3-chloro-2-fluoropyridine,,pyridine,5-bromo-3-chloro-2-fluoro,pyridine, 5-bromo-3-chloro-2-fluoro
12056904
5-bromo-3-chloro-2-fluoropyridine
C1=C(C(=NC=C1Br)F)Cl
38185-56-7
C5H2BrClFN
210.43
MFCD09834365
WXKLHBHJIBAXQH-UHFFFAOYSA-N
2-fluoro-3-chloro-5-bromopyridine,3-bromo-5-chloro-6-fluoropyridine,5-bromo-3-chloro-2-fluoro-pyridine,acmc-1ct80,5-bromo-3-chloro-2-fluoropyridine,,pyridine,5-bromo-3-chloro-2-fluoro,pyridine, 5-bromo-3-chloro-2-fluoro
12056904
5-bromo-3-chloro-2-fluoropyridine
C1=C(C(=NC=C1Br)F)Cl
Quantity 250mg
Melting Point 60°C
Beilstein 1563886
Formula Weight 210.43
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 5-Bromo-3-chloro-2-fluoropyridine

storageNote1 : 2° to 8°C