Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromo-3-chloro-2-hydroxypyridine, 97%

Click to view available options
Quantity:
250mg
1g
5g
58236-70-7
C5H3BrClNO
208.439
MFCD05662389
IPWYUROVLMVBGD-UHFFFAOYSA-N
5-bromo-3-chloropyridin-2-ol,5-bromo-3-chloro-2-hydroxypyridine,5-bromo-3-chloropyridin-2 1h-one,5-bromo-3-chloro-2-pyridinone,5-bromo-3-chloro-2-pyridone,2 1h-pyridinone, 5-bromo-3-chloro,5-bromo-3-chloro-1,2-dihydropyridin-2-one,5-bromo-2-hydroxy-3-chloropyridine,pubchem1172,ksc608a4r
19050009
5-bromo-3-chloro-1H-pyridin-2-one
C1=C(C(=O)NC=C1Br)Cl
58236-70-7
C5H3BrClNO
208.439
MFCD05662389
IPWYUROVLMVBGD-UHFFFAOYSA-N
5-bromo-3-chloropyridin-2-ol,5-bromo-3-chloro-2-hydroxypyridine,5-bromo-3-chloropyridin-2 1h-one,5-bromo-3-chloro-2-pyridinone,5-bromo-3-chloro-2-pyridone,2 1h-pyridinone, 5-bromo-3-chloro,5-bromo-3-chloro-1,2-dihydropyridin-2-one,5-bromo-2-hydroxy-3-chloropyridine,pubchem1172,ksc608a4r
19050009
5-bromo-3-chloro-1H-pyridin-2-one
C1=C(C(=O)NC=C1Br)Cl
Quantity 250mg
Melting Point 199°C to 202°C
Beilstein 1448359
Formula Weight 208.44
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-chloro-2-hydroxypyridine