Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-3-fluoropyridine-2-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
669066-91-5
C6H3BrFNO2
220
JXHUXOAJAYWMHK-UHFFFAOYSA-N
5-bromo-3-fluoropicolinic acid,5-bromo-3-fluoro-2-pyridinecarboxylic acid,2-pyridinecarboxylic acid, 5-bromo-3-fluoro,5-bromo-3-fluoropicolinicacid,5-bromo-2-carboxy-3-fluoropyridine,5-bromo-3-fluoropyridin-2-carboxylicacid,2-pyridinecarboxylicacid,5-bromo-3-fluoro,2-pyridinecarboxylicacid, 5-bromo-3-fluoro,5-bromo-3-fluoro-pyridine-2-carboxylic acid,5-bromanyl-3-fluoranyl-pyridine-2-carboxylic acid
21075872
5-bromo-3-fluoropyridine-2-carboxylic acid
C1=C(C(=NC=C1Br)C(=O)O)F
669066-91-5
C6H3BrFNO2
220
JXHUXOAJAYWMHK-UHFFFAOYSA-N
5-bromo-3-fluoropicolinic acid,5-bromo-3-fluoro-2-pyridinecarboxylic acid,2-pyridinecarboxylic acid, 5-bromo-3-fluoro,5-bromo-3-fluoropicolinicacid,5-bromo-2-carboxy-3-fluoropyridine,5-bromo-3-fluoropyridin-2-carboxylicacid,2-pyridinecarboxylicacid,5-bromo-3-fluoro,2-pyridinecarboxylicacid, 5-bromo-3-fluoro,5-bromo-3-fluoro-pyridine-2-carboxylic acid,5-bromanyl-3-fluoranyl-pyridine-2-carboxylic acid
21075872
5-bromo-3-fluoropyridine-2-carboxylic acid
C1=C(C(=NC=C1Br)C(=O)O)F
Packaging Glass Bottle
Melting Point 165°C to 170°C
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 220
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-fluoropyridine-2-carboxylic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning