Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-GAL), >99%, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
100mg
250mg
1g
2g
5g
Packaging:
Plastic bottle
7240-90-6
C14H15BrClNO6
408.629
MFCD00005666
OPIFSICVWOWJMJ-AEOCFKNESA-N
x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal
65181
CHEBI:75055
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
7240-90-6
C14H15BrClNO6
408.629
MFCD00005666
OPIFSICVWOWJMJ-AEOCFKNESA-N
x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal
65181
CHEBI:75055
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Histochemical substrate for β-Galactosidase and substrate for blue/white colony screening

A histochemical substrate for β-Galactosidase. A substrate for blue/white colony screening. Effective concentration is approximately 50 μg/ml.
Melting Point ~236°C (decomposition)
Packaging Plastic bottle
Quantity 250mg
Formula Weight 408.63
Physical Form Crystalline Powder
Percent Purity ≥99%
Grade Ultrapure
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-GAL)