Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-GAL), >99%, Ultrapure, Thermo Scientific™

Catalog Number AAJ1007703
Click to view available options
Quantity:
100 mg
250 mg
1 g
Packaging:
Plastic bottle
7240-90-6
C14H15BrClNO6
408.629
MFCD00005666
OPIFSICVWOWJMJ-AEOCFKNESA-N
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
65181
CHEBI:75055
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
7240-90-6
C14H15BrClNO6
408.629
MFCD00005666
OPIFSICVWOWJMJ-AEOCFKNESA-N
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
65181
CHEBI:75055
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Histochemical substrate for β-Galactosidase and substrate for blue/white colony screening

A histochemical substrate for β-Galactosidase. A substrate for blue/white colony screening. Effective concentration is approximately 50 μg/ml.
TRUSTED_SUSTAINABILITY
Melting Point ∼236°C (decomposition)
Beilstein 1402009
Packaging Plastic bottle
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 408.63
Percent Purity ≥99%
Grade Ultrapure
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-GAL)
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 230-640-8
TSCA No
Recommended Storage Keep cold
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit