Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-GAL), >99%, Ultrapure, Thermo Scientific™
Click to view available options
Quantity:
100 mg
250 mg
1 g
Packaging:
Plastic bottle
Description
A histochemical substrate for β-Galactosidase. A substrate for blue/white colony screening. Effective concentration is approximately 50 μg/ml.
Specifications
Melting Point | ∼236°C (decomposition) |
Beilstein | 1402009 |
Packaging | Plastic bottle |
Quantity | 100 mg |
Sensitivity | Light sensitive |
Formula Weight | 408.63 |
Percent Purity | ≥99% |
Grade | Ultrapure |
Physical Form | Crystalline Powder |
Chemical Name or Material | 5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-GAL) |
Safety and Handling
Hazard Category | H302+H312+H332 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
EINECSNumber | 230-640-8 |
TSCA | No |
Recommended Storage | Keep cold |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction