Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-chloro-3-indolyl sulfate potassium salt, 99%, ACROS Organics™

Click to view available options
Quantity:
200mg
Packaging:
Glass bottle
6578-07-0
C8H4BrClKNO4S
364.635
MFCD00056929
BONBCMDYPZGTEU-UHFFFAOYSA-M
5-bromo-4-chloro-3-indolyl sulfate potassium salt,potassium 5-bromo-4-chloro-1h-indol-3-yl sulfate,5-bromo-4-chloro-3-indoxyl sulfate, potassium salt,c8h4brclno4s.k,5-bromo-4-chloro-1h-indol-3-yl sulfate potassium salt,5-bromo-4-chloroindol-3-yl hydroxysulfonate, potassium salt,5-bromo-4-chloro-3-indolyl sulfate potassium salt, sulfatase substrate,x-sulfate potassium salt;sulfuric acid-mono-5-bromo-4-chloro-1h-indol-3-yl ester potassium salt
23664775
potassium;(5-bromo-4-chloro-1H-indol-3-yl) sulfate
C1=CC(=C(C2=C1NC=C2OS(=O)(=O)[O-])Cl)Br.[K+]
6578-07-0
C8H4BrClKNO4S
364.635
MFCD00056929
BONBCMDYPZGTEU-UHFFFAOYSA-M
5-bromo-4-chloro-3-indolyl sulfate potassium salt,potassium 5-bromo-4-chloro-1h-indol-3-yl sulfate,5-bromo-4-chloro-3-indoxyl sulfate, potassium salt,c8h4brclno4s.k,5-bromo-4-chloro-1h-indol-3-yl sulfate potassium salt,5-bromo-4-chloroindol-3-yl hydroxysulfonate, potassium salt,5-bromo-4-chloro-3-indolyl sulfate potassium salt, sulfatase substrate,x-sulfate potassium salt;sulfuric acid-mono-5-bromo-4-chloro-1h-indol-3-yl ester potassium salt
23664775
potassium;(5-bromo-4-chloro-1H-indol-3-yl) sulfate
C1=CC(=C(C2=C1NC=C2OS(=O)(=O)[O-])Cl)Br.[K+]
Infrared Spectrum Authentic
Color White to Yellow
Quantity 200mg
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 364.63
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl sulfate potassium salt, 99%
Show More Show Less