Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-chloro-3-indoxyl-β-D-xylopyranoside, 99%, Acros Organics

Click to view available options
Quantity:
100mg
Packaging:
Glass bottle
207606-55-1
C13H13BrClNO5
378.603
MFCD00152013
LXVCKAHHHVVOJV-KQXJFYCDSA-N
5-bromo-4-chloro-3-indoxyl-beta-d-xylopyranoside,x-xyl,5-bromo-4-chloro-3-indolyl beta-d-xylopyranoside,2s,3r,4s,5r-2-5-bromo-4-chloro-1h-indol-3-yl oxy tetrahydro-2h-pyran-3,4,5-triol,5-bromo-4-chloro-3-indolyl,a-d-xylopyranoside,5-bromo-4-chloro-3-indolyl-beta-d-xylopyranoside,2s,3r,4s,5r-2-5-bromo-4-chloro-1h-indol-3-yl oxy oxane-3,4,5-triol,5-bromo-4-chloro-3-indolyl b-d-xylopyranoside
2733917
(2S,3R,4S,5R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]oxane-3,4,5-triol
C1C(C(C(C(O1)OC2=CNC3=C2C(=C(C=C3)Br)Cl)O)O)O
207606-55-1
C13H13BrClNO5
378.603
MFCD00152013
LXVCKAHHHVVOJV-KQXJFYCDSA-N
5-bromo-4-chloro-3-indoxyl-beta-d-xylopyranoside,x-xyl,5-bromo-4-chloro-3-indolyl beta-d-xylopyranoside,2s,3r,4s,5r-2-5-bromo-4-chloro-1h-indol-3-yl oxy tetrahydro-2h-pyran-3,4,5-triol,5-bromo-4-chloro-3-indolyl,a-d-xylopyranoside,5-bromo-4-chloro-3-indolyl-beta-d-xylopyranoside,2s,3r,4s,5r-2-5-bromo-4-chloro-1h-indol-3-yl oxy oxane-3,4,5-triol,5-bromo-4-chloro-3-indolyl b-d-xylopyranoside
2733917
(2S,3R,4S,5R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]oxane-3,4,5-triol
C1C(C(C(C(O1)OC2=CNC3=C2C(=C(C=C3)Br)Cl)O)O)O
Specific Rotation -90° to -94° (20°C, 589nm) (c=0.5, MeOH)
Quantity 100mg
Infrared Spectrum Authentic
Color White
Water 1% Max. (K.F.)
Packaging Glass bottle
Appearance (c=1, MeOH) clear, colorless
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 378.61
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 5-Bromo-4-chloro-3-indoxyl-β-D-xylopyranoside
Show More Show Less

Recommended Storage : Freezer -20°C