Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-6-chloro-2-picoline, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
185017-72-5
C6H5BrClN
206.5
MFCD03095218
JVDQYSIJBRTRMS-UHFFFAOYSA-N
3-bromo-2-chloro-6-picoline,5-bromo-6-chloro-2-picoline,2-chloro-3-bromo-6-methylpyridine,5-bromo-6-chloro-2-methylpyridine,pyridine, 3-bromo-2-chloro-6-methyl,3-bromo-2-chloro-6-methyl-pyridine,pubchem1286,acmc-209emg,ksc538q8t,3-bromo-2-chloro-6 picoline
2734418
3-bromo-2-chloro-6-methylpyridine
CC1=NC(=C(C=C1)Br)Cl
185017-72-5
C6H5BrClN
206.5
MFCD03095218
JVDQYSIJBRTRMS-UHFFFAOYSA-N
3-bromo-2-chloro-6-picoline,5-bromo-6-chloro-2-picoline,2-chloro-3-bromo-6-methylpyridine,5-bromo-6-chloro-2-methylpyridine,pyridine, 3-bromo-2-chloro-6-methyl,3-bromo-2-chloro-6-methyl-pyridine,pubchem1286,acmc-209emg,ksc538q8t,3-bromo-2-chloro-6 picoline
2734418
3-bromo-2-chloro-6-methylpyridine
CC1=NC(=C(C=C1)Br)Cl
Quantity 1g
Packaging Glass bottle
Color Yellow
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Formula Weight 206.5
Physical Form Low Melting Solid or Liquid
Percent Purity 98+%
Chemical Name or Material 5-Bromo-6-chloro-2-picoline
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning