Alfa Aesar™ 5-Bromoacetyl-2-hydroxybenzamide, 95%

Manufacturer: Alfa Aesar™ B2433018

Quantity 50g

 View more versions of this product

Catalog No. AAB2433018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 73866-23-6
    Molecular Formula C9H8BrNO3
    Formula Weight 258.07
    MDL Number MFCD00792456
    Synonym 5-(Bromoacetyl)salicylamide; 2-Bromo-3'-carboxamido-4'-hydroxyacetophenone
    Chemical Name or Material 5-Bromoacetyl-2-hydroxybenzamide

    Specifications

    Formula Weight 258.07
    Molecular Formula C9H8BrNO3
    MDL Number MFCD00792456
    Melting Point 202° to 204°C
    Synonym 5-(Bromoacetyl)salicylamide; 2-Bromo-3'-carboxamido-4'-hydroxyacetophenone
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number CV2365900
    CAS 73866-23-6
    Health Hazard 1 Warning
    Chemical Name or Material 5-Bromoacetyl-2-hydroxybenzamide
    Assay 95%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit