Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromoamyl acetate, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
Packaging:
Glass bottle
15848-22-3
C7H13BrO2
209.083
MFCD00039204
JIHJLQWUVZUKCH-UHFFFAOYSA-N
5-bromoamyl acetate,5-bromo-n-amyl acetate,1-acetoxy-5-bromopentane,1-pentanol, 5-bromo-,1-acetate,5-?bromopentyl acetate,acmc-209dhw,5-bromopentylacetate,5-bromo-1-pentanyl acetate,5-bromopentyl acetate,acetic acid 5-bromoamyl ester
85140
5-bromopentyl acetate
CC(=O)OCCCCCBr
15848-22-3
C7H13BrO2
209.083
MFCD00039204
JIHJLQWUVZUKCH-UHFFFAOYSA-N
5-bromoamyl acetate,5-bromo-n-amyl acetate,1-acetoxy-5-bromopentane,1-pentanol, 5-bromo-,1-acetate,5-?bromopentyl acetate,acmc-209dhw,5-bromopentylacetate,5-bromo-1-pentanyl acetate,5-bromopentyl acetate,acetic acid 5-bromoamyl ester
85140
5-bromopentyl acetate
CC(=O)OCCCCCBr
Quantity 5mL
Refractive Index 1.4600 to 1.4620 (20°C, 589nm)
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3COO(CH)5Br
Formula Weight 209.08
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 5-Bromoamyl acetate
Show More Show Less