Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromobenzo[b]furan, 97%, Acros Organics™

Click to view available options
Quantity:
1g
23145-07-5
C8H5BrO
197.03
AYOVPQORFBWFNO-UHFFFAOYSA-N
5-bromobenzofuran,benzofuran, 5-bromo,5-bromobenzo b furan,5-bromo-benzofuran,5-bromobenzo beta furan,buttpark 95\50-66,pubchem22017,5-bromobenzo b-furan,#
90015
5-bromo-1-benzofuran
C1=CC2=C(C=CO2)C=C1Br
23145-07-5
C8H5BrO
197.03
AYOVPQORFBWFNO-UHFFFAOYSA-N
5-bromobenzofuran,benzofuran, 5-bromo,5-bromobenzo b furan,5-bromo-benzofuran,5-bromobenzo beta furan,buttpark 95\50-66,pubchem22017,5-bromobenzo b-furan,#
90015
5-bromo-1-benzofuran
C1=CC2=C(C=CO2)C=C1Br
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 5-Bromobenzo[b]furan

GHS H Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning