Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 5-Bromoindole-3-carboxylic acid, 97%
Click to view available options
Quantity:
250mg
1g
5g
Specifications
Quantity | 5g |
Beilstein | 475219 |
Formula Weight | 240.06 |
Physical Form | Solid |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromoindole-3-carboxylic acid |
Safety and Handling
storageNote1 : 2° to 8°C