Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromoindole-3-carboxylic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
10406-06-1
C9H6BrNO2
240.056
MFCD05664007
JVZMBSGNSAHFCY-UHFFFAOYSA-N
5-bromoindole-3-carboxylic acid,5-bromo-indole-3-carboxylic acid,5-bromo-3-indolecarboxylic acid,1h-indole-3-carboxylic acid,5-bromo,1h-indole-3-carboxylic acid, 5-bromo,5-bromo-1h-indole-3-carboxylicacid,pubchem7982,acmc-1btmm,ksc173q4r,5-bromo-3-indolecarboxylicacid
7018243
5-bromo-1H-indole-3-carboxylic acid
C1=CC2=C(C=C1Br)C(=CN2)C(=O)O
10406-06-1
C9H6BrNO2
240.056
MFCD05664007
JVZMBSGNSAHFCY-UHFFFAOYSA-N
5-bromoindole-3-carboxylic acid,5-bromo-indole-3-carboxylic acid,5-bromo-3-indolecarboxylic acid,1h-indole-3-carboxylic acid,5-bromo,1h-indole-3-carboxylic acid, 5-bromo,5-bromo-1h-indole-3-carboxylicacid,pubchem7982,acmc-1btmm,ksc173q4r,5-bromo-3-indolecarboxylicacid
7018243
5-bromo-1H-indole-3-carboxylic acid
C1=CC2=C(C=C1Br)C(=CN2)C(=O)O
Quantity 250mg
Beilstein 475219
Formula Weight 240.06
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 5-Bromoindole-3-carboxylic acid

storageNote1 : 2° to 8°C