Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromoindoxyl diacetate, 98+%

Click to view available options
Quantity:
250mg
1g
33588-54-4
C12H10BrNO3
296.12
MFCD00005799
XJRIDJAGAYGJCK-UHFFFAOYSA-N
5-bromoindoxyl diacetate,1-acetyl-5-bromo-1h-indol-3-yl acetate,5-bromoindoxyldiacetate,n-acetyl-5-bromoindolyl acetate,1-acetyl-3-acetyloxy-5-bromoindole,3-acetoxy-1-acetyl-5-bromoindole,1h-indol-3-ol, 1-acetyl-5-bromo-, acetate ester,3-acetoxy-1-acetyl-5-bromo-indole,1h-indol-3-ol, 1-acetyl-5-bromo-, acetate,pubchem16263
118505
(1-acetyl-5-bromoindol-3-yl) acetate
CC(=O)N1C=C(C2=C1C=CC(=C2)Br)OC(=O)C
33588-54-4
C12H10BrNO3
296.12
MFCD00005799
XJRIDJAGAYGJCK-UHFFFAOYSA-N
5-bromoindoxyl diacetate,1-acetyl-5-bromo-1h-indol-3-yl acetate,5-bromoindoxyldiacetate,n-acetyl-5-bromoindolyl acetate,1-acetyl-3-acetyloxy-5-bromoindole,3-acetoxy-1-acetyl-5-bromoindole,1h-indol-3-ol, 1-acetyl-5-bromo-, acetate ester,3-acetoxy-1-acetyl-5-bromo-indole,1h-indol-3-ol, 1-acetyl-5-bromo-, acetate,pubchem16263
118505
(1-acetyl-5-bromoindol-3-yl) acetate
CC(=O)N1C=C(C2=C1C=CC(=C2)Br)OC(=O)C
Melting Point 122°C to 125°C
Quantity 250mg
Beilstein 236935
Formula Weight 296.13
Percent Purity ≥98%
Chemical Name or Material 5-Bromoindoxyl diacetate

EINECSNumber : 251-584-0

TSCA : No

Recommended Storage : Keep cold