Alfa Aesar™ 5-Bromoisophthalic acid, 95%

Manufacturer: Alfa Aesar™ H2687414

Quantity 25g

 View more versions of this product

Catalog No. AAH2687414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 23351-91-9
    Molecular Formula C8H5BrO4
    Formula Weight 245.03
    MDL Number MFCD00152019
    Chemical Name or Material 5-Bromoisophthalic acid

    Specifications

    Formula Weight 245.03
    TSCA TSCA
    Molecular Formula C8H5BrO4
    MDL Number MFCD00152019
    CAS 23351-91-9
    Melting Point 280° to 286°C
    Quantity 25g
    Odor Odorless
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 5-Bromoisophthalic acid
    Assay 95%

    Description

    Applications
    5-Bromoisophthalic acid, is used as a pharmaceutical intermediate. It is also used in the polymer industry.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit